2-Quinoxalinol 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD75905-1000GEA 1520 DKK
BLDPBD75905-1000G BLDPBD75905-100G BLDPBD75905-10G BLDPBD75905-500G BLDPBD75905-0.5G BLDPBD75905-25G
2-Quinoxalinol 98%
2-Quinoxalinol

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₆N₂O
Molekylvægt: 146,15 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006722
CAS-nummer: 1196-57-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR