Isoquinoline-5-sulphonic acid 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD7483-5GEA 54.3 DKK
BLDPBD7483-5G BLDPBD7483-25G BLDPBD7483-10G
Isoquinoline-5-sulphonic acid 95%
Isoquinoline-5-sulphonic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₇NO₃S
Molekylvægt: 209,22 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00134089
CAS-nummer: 27655-40-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR