5-Methylisatoic anhydride 96%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD7482-25GEA 2280 DKK
BLDPBD7482-25G BLDPBD7482-250MG BLDPBD7482-0.025G BLDPBD7482-10G BLDPBD7482-5G BLDPBD7482-1G
5-Methylisatoic anhydride 96%
5-Methylisatoic anhydride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₉H₇NO₃
Molekylvægt: 177,1568 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06809783
CAS-nummer: 4692-99-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR