2'-Cyanoacetanilide 97%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD74090-5GEA
311
DKK
BLDPBD74090-5G
BLDPBD74090-1G
2'-Cyanoacetanilide 97%
2'-Cyanoacetanilide
Storage condition: Sealed in dry, Room Temperature
Formel:
C₉H₈N₂O Molekylvægt: 160,1726 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00019840 CAS-nummer: 621-03-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...