3-Amino-1-bromoisoquinoline 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD7355-1GEA 845 DKK
BLDPBD7355-1G BLDPBD7355-250MG BLDPBD7355-5G BLDPBD7355-100MG
3-Amino-1-bromoisoquinoline 98%
3-Amino-1-bromoisoquinoline

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₉H₇BrN₂
Molekylvægt: 223,07 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00102193
CAS-nummer: 13130-79-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR