2-Chloro-5-iodo-3-methylpyridine 97%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD73194-1GEA 295 DKK
BLDPBD73194-1G BLDPBD73194-5G BLDPBD73194-10G
2-Chloro-5-iodo-3-methylpyridine 97%
2-Chloro-5-iodo-3-methylpyridine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₆H₅ClIN
Molekylvægt: 253,468 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD09037455
CAS-nummer: 59782-89-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR