2-Bromopropanamide 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD72285-100GEA 5470 DKK
BLDPBD72285-100G BLDPBD72285-5G BLDPBD72285-1G BLDPBD72285-250MG BLDPBD72285-25G
2-Bromopropanamide 97%
2-Bromopropanamide

Storage condition: Inert atmosphere, Room Temperature

Formel: C₃H₆BrNO
Molekylvægt: 151,99 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00012376
CAS-nummer: 5875-25-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR