2-Bromo-4'-(trifluoromethoxy)acetophenone 97%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD7213-10GEA
388
DKK
BLDPBD7213-10G
BLDPBD7213-1G
BLDPBD7213-5G
BLDPBD7213-250MG
BLDPBD7213-25G
2-Bromo-4'-(trifluoromethoxy)acetophenone 97%
2-Bromo-4'-(trifluoromethoxy)acetophenone
Storage condition: Inert atmosphere, 2-8°C
Formel:
C₉H₆BrF₃O₂ Molekylvægt: 283,04 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00052333 CAS-nummer: 103962-10-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...