(±)-1-Boc-3-aminopyrrolidine 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD7114-5GEA 225 DKK
BLDPBD7114-5G BLDPBD7114-1G BLDPBD7114-25G BLDPBD7114-10G
(±)-1-Boc-3-aminopyrrolidine 97%
(±)-1-Boc-3-aminopyrrolidine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₉H₁₈N₂O₂
Molekylvægt: 186,25 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01861220
CAS-nummer: 186550-13-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR