2-Amino-6-chloro-4-nitrophenol 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD70594-5GEA 326 DKK
BLDPBD70594-0.025G BLDPBD70594-5G BLDPBD70594-1G BLDPBD70594-10G BLDPBD70594-25G
2-Amino-6-chloro-4-nitrophenol 97%
2-Amino-6-chloro-4-nitrophenol

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₆H₅ClN₂O₃
Molekylvægt: 188,57 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00035767
CAS-nummer: 6358-09-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR