2-Amino-5-bromo-4-methyl-3-nitropyridine 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD70356-5GEA 241 DKK
BLDPBD70356-5G BLDPBD70356-10G BLDPBD70356-250MG BLDPBD70356-1G
2-Amino-5-bromo-4-methyl-3-nitropyridine 95%
2-Amino-5-bromo-4-methyl-3-nitropyridine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₆H₆BrN₃O₂
Molekylvægt: 232,0400 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00092197
CAS-nummer: 100367-40-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR