Guanine hydrochloride 98%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BLDPBD69620-5GEA
62.1
DKK
BLDPBD69620-5G
BLDPBD69620-25G
BLDPBD69620-10G
BLDPBD69620-100G
Guanine hydrochloride 98%
Guanine hydrochloride
Formel:
C₅H₆ClN₅O Molekylvægt: 187,59 g/mol |
MDL Number:
MFCD00213670 CAS-nummer: 635-39-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...