4-Amino-N,N-dimethylbenzamide 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD6894-100MGEA 171 DKK
BLDPBD6894-100MG BLDPBD6894-250MG BLDPBD6894-1G BLDPBD6894-10G BLDPBD6894-5G
4-Amino-N,N-dimethylbenzamide 97%
4-Amino-N,N-dimethylbenzamide

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₉H₁₂N₂O
Molekylvægt: 164,21 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00523648
CAS-nummer: 6331-71-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR