4-(2-Pyrrolidinyl)pyridine 97%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD6887-1GEA
752
DKK
BLDPBD6887-1G
BLDPBD6887-250MG
BLDPBD6887-5G
4-(2-Pyrrolidinyl)pyridine 97%
4-(2-Pyrrolidinyl)pyridine
Storage condition: Keep in dark place, Inert atmosphere, 2-8°C
Formel:
C₉H₁₂N₂ Molekylvægt: 148,21 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD01862536 CAS-nummer: 128562-25-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...