2,6-Dichloroisonicotinamide 95%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD68184-5GEA 969 DKK
BLDPBD68184-5G BLDPBD68184-10G BLDPBD68184-25G BLDPBD68184-1G
2,6-Dichloroisonicotinamide 95%
2,6-Dichloroisonicotinamide

Storage condition: Inert atmosphere, Room Temperature

Formel: C₆H₄Cl₂N₂O
Molekylvægt: 191,0200 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00052640
CAS-nummer: 89281-13-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR