2'-Amino-4'-methoxyacetophenone 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD6816-250MGEA 853 DKK
BLDPBD6816-250MG BLDPBD6816-1G BLDPBD6816-100MG BLDPBD6816-5G
2'-Amino-4'-methoxyacetophenone 98%
2'-Amino-4'-methoxyacetophenone

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₉H₁₁NO₂
Molekylvægt: 165,1900 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD04972112
CAS-nummer: 42465-53-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR