Methyl-2-amino-3-methylbenzoate 96%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD6809-100GEA 861 DKK
BLDPBD6809-100G BLDPBD6809-5G BLDPBD6809-1G BLDPBD6809-10G BLDPBD6809-25G
Methyl-2-amino-3-methylbenzoate 96%
Methyl-2-amino-3-methylbenzoate

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₉H₁₁NO₂
Molekylvægt: 165,19 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06200918
CAS-nummer: 22223-49-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR