2,5-Dimethyl-1H-benzimidazole 95+%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD67755-250MGEA 125 DKK
BLDPBD67755-250MG BLDPBD67755-1G BLDPBD67755-5G
2,5-Dimethyl-1H-benzimidazole 95+%
2,5-Dimethyl-1H-benzimidazole

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₁₀N₂
Molekylvægt: 146,189 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00125906
CAS-nummer: 1792-41-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR