2',5'-Difluoroacetanilide 97%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD67608-10GEA
621
DKK
BLDPBD67608-10G
BLDPBD67608-5G
BLDPBD67608-25G
BLDPBD67608-1G
2',5'-Difluoroacetanilide 97%
2',5'-Difluoroacetanilide
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formel:
C₈H₇F₂NO Molekylvægt: 171,1441 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00032512 CAS-nummer: 398-90-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...