3-Amino-3-(2-nitrophenyl)propionic acid 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD6637-5GEA 1730 DKK
BLDPBD6637-5G BLDPBD6637-1G BLDPBD6637-250MG
3-Amino-3-(2-nitrophenyl)propionic acid 98%
3-Amino-3-(2-nitrophenyl)propionic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₉H₁₀N₂O₄
Molekylvægt: 210,19 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00090356
CAS-nummer: 5678-48-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR