2,3-Dihydro-1,4-benzodioxine-5-carboxamide 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD65613-1GEA 1160 DKK
BLDPBD65613-1G BLDPBD65613-100MG BLDPBD65613-250MG
2,3-Dihydro-1,4-benzodioxine-5-carboxamide 95%
2,3-Dihydro-1,4-benzodioxine-5-carboxamide

Storage condition: Sealed in dry, 2-8°C

Formel: C₉H₉NO₃
Molekylvægt: 179,173 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD11053918
CAS-nummer: 349550-81-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR