D-(-)-α-(4-Hydroxyphenyl)glycine 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD6550-10GEA 46.6 DKK
BLDPBD6550-10G BLDPBD6550-100G BLDPBD6550-25G BLDPBD6550-500G
D-(-)-α-(4-Hydroxyphenyl)glycine 98%
D-(-)-α-(4-Hydroxyphenyl)glycine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₈H₉NO₃
Molekylvægt: 167,16 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00004262
CAS-nummer: 22818-40-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR