Methyl-2,2-dimethoxypropionate 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD64486-1GEA 163 DKK
BLDPBD64486-1G BLDPBD64486-25G BLDPBD64486-5G BLDPBD64486-10G
Methyl-2,2-dimethoxypropionate 98%
Methyl-2,2-dimethoxypropionate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₆H₁₂O₄
Molekylvægt: 148,1571 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00048066
CAS-nummer: 10076-48-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR