2,1,3-Benzothiadiazol-5-ylamine 97%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD63766-250MGEA 125 DKK
BLDPBD63766-250MG BLDPBD63766-1G
2,1,3-Benzothiadiazol-5-ylamine 97%
2,1,3-Benzothiadiazol-5-ylamine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₆H₅N₃S
Molekylvægt: 151,1900 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00185826
CAS-nummer: 874-37-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR