Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')cobalt(III) 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD634522-5GEA 3620 DKK
BLDPBD634522-5G BLDPBD634522-1G BLDPBD634522-250MG
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')cobalt(III) 98%
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')cobalt(III)

Storage condition: Inert atmosphere, 2-8°C

Formel: C₃₃H₅₇CoO₆
Molekylvægt: 608,735 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00233617
CAS-nummer: 14877-41-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR