2-(6-Chloro-3-pyridazinyl)-2-phenylacetonitrile 95%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD62745-1GEA
969
DKK
BLDPBD62745-1G
BLDPBD62745-250MG
BLDPBD62745-5G
2-(6-Chloro-3-pyridazinyl)-2-phenylacetonitrile 95%
2-(6-Chloro-3-pyridazinyl)-2-phenylacetonitrile
Storage condition: Inert atmosphere, 2-8°C
Formel:
C₁₂H₈ClN₃ Molekylvægt: 229,6650 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00140699 CAS-nummer: 73535-73-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...