3-Fluoro-(5-methoxycarbonyl)phenylboronic acid 98%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD6256-100MGEA
132
DKK
BLDPBD6256-100MG
BLDPBD6256-250MG
BLDPBD6256-1G
3-Fluoro-(5-methoxycarbonyl)phenylboronic acid 98%
3-Fluoro-(5-methoxycarbonyl)phenylboronic acid
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₈H₈BFO₄ Molekylvægt: 197,96 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD07363749 CAS-nummer: 871329-62-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...