5-Methoxy-2-nitrobenzaldehyde 98%
Leverandør: BLD PHARMATECH GMBH
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD6234-25GEA
1420
DKK
BLDPBD6234-25G
BLDPBD6234-10G
BLDPBD6234-100G
BLDPBD6234-250MG
BLDPBD6234-1G
BLDPBD6234-5G
5-Methoxy-2-nitrobenzaldehyde 98%
5-Methoxy-2-nitrobenzaldehyde
Storage condition: Inert atmosphere, 2-8°C
Formel:
C₈H₇NO₄ Molekylvægt: 181,1500 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00456498 CAS-nummer: 20357-24-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...