2-Chloro-4-(methylsulphonyl)benzoic acid 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD6120-25GEA 69.8 DKK
BLDPBD6120-25G BLDPBD6120-100G BLDPBD6120-5G
2-Chloro-4-(methylsulphonyl)benzoic acid 98%
2-Chloro-4-(methylsulphonyl)benzoic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₇ClO₄S
Molekylvægt: 234,66 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00216496
CAS-nummer: 53250-83-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR