2-Amino-5-bromobenzoylpyridine 98%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD61051-100GEA
2930
DKK
BLDPBD61051-100G
BLDPBD61051-5G
BLDPBD61051-1G
BLDPBD61051-10G
BLDPBD61051-25G
2-Amino-5-bromobenzoylpyridine 98%
2-Amino-5-bromobenzoylpyridine
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formel:
C₁₂H₉BrN₂O Molekylvægt: 277,1167 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD07780283 CAS-nummer: 1563-56-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...