Ethyl-1H-indazole-3-carboxylate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD58791-250MGEA 54.3 DKK
BLDPBD58791-250MG BLDPBD58791-5G BLDPBD58791-1G BLDPBD58791-25G BLDPBD58791-10G BLDPBD58791-100MG
Ethyl-1H-indazole-3-carboxylate 95%
Ethyl-1H-indazole-3-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₁₀N₂O₂
Molekylvægt: 190,20 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01138134
CAS-nummer: 4498-68-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR