1-Chloro-2-(trifluoromethoxy)benzene 98%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD57715-100GEA 458 DKK
BLDPBD57715-100G BLDPBD57715-25G BLDPBD57715-500G BLDPBD57715-5G BLDPBD57715-10G
1-Chloro-2-(trifluoromethoxy)benzene 98%
1-Chloro-2-(trifluoromethoxy)benzene

Storage condition: Sealed in dry, Room Temperature

Formel: C₇H₄ClF₃O
Molekylvægt: 196,5543 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD03840206
CAS-nummer: 450-96-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR