4-(Difluoromethoxy)benzonitrile 98%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD5729-5GEA
458
DKK
BLDPBD5729-5G
BLDPBD5729-1G
4-(Difluoromethoxy)benzonitrile 98%
4-(Difluoromethoxy)benzonitrile
Storage condition: Inert atmosphere, 2-8°C
Formel:
C₈H₅F₂NO Molekylvægt: 169,1282 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00085006 CAS-nummer: 90446-25-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...