1-Bromo-2,4,6-tri-tert-butylbenzene 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD57285-1GEA 187 DKK
BLDPBD57285-1G BLDPBD57285-10G BLDPBD57285-100MG BLDPBD57285-100G BLDPBD57285-5G BLDPBD57285-25G BLDPBD57285-250MG
1-Bromo-2,4,6-tri-tert-butylbenzene 97%
1-Bromo-2,4,6-tri-tert-butylbenzene

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₈H₂₉Br
Molekylvægt: 325,3300 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00191861
CAS-nummer: 3975-77-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR