6-Bromo-8-methoxy-[1,2,4]triazolo[4,3-a]pyrazine 97%
Leverandør: BLD PHARMATECH GMBH
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD562096-250MGEA
2760
DKK
BLDPBD562096-250MG
BLDPBD562096-1G
BLDPBD562096-100MG
6-Bromo-8-methoxy-[1,2,4]triazolo[4,3-a]pyrazine 97%
6-Bromo-8-methoxy-[1,2,4]triazolo[4,3-a]pyrazine
Storage condition: Inert atmosphere, 2-8°C
Formel:
C₆H₅BrN₄O Molekylvægt: 229,034 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD20501877 CAS-nummer: 1350885-67-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...