10-Hydroxydecanoic acid 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD55722-10GEA 241 DKK
BLDPBD55722-10G BLDPBD55722-5G BLDPBD55722-500G BLDPBD55722-1000G BLDPBD55722-100G BLDPBD55722-1G BLDPBD55722-25G BLDPBD55722-0.5G
10-Hydroxydecanoic acid 98%
10-Hydroxydecanoic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₂₀O₃
Molekylvægt: 188,26 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010510
CAS-nummer: 1679-53-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR