Dinatrium naphthalen-1,5-disulphonat 98%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD54983-1000GEA
311
DKK
BLDPBD54983-1000G
BLDPBD54983-25G
BLDPBD54983-500G
BLDPBD54983-0.025G
BLDPBD54983-100G
Dinatrium naphthalen-1,5-disulphonat 98%
Dinatrium naphthalen-1,5-disulphonat
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₁₀H₆Na₂O₆S₂ Molekylvægt: 332,27 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00064178 CAS-nummer: 1655-29-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...