1,4-Benzodioxane-6-boronic acid pinacol ester 98%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD54412-1GEA
380
DKK
BLDPBD54412-1G
BLDPBD54412-100MG
BLDPBD54412-250MG
1,4-Benzodioxane-6-boronic acid pinacol ester 98%
1,4-Benzodioxane-6-boronic acid pinacol ester
Storage condition: Sealed in dry, 2-8°C
Formel:
C₁₄H₁₉BO₄ Molekylvægt: 262,1093 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD05663890 CAS-nummer: 517874-21-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...