trans-1,4-Cyclohexanedicarboxylic acid 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD5394-25GEA 85.3 DKK
BLDPBD5394-25G BLDPBD5394-500G BLDPBD5394-10G BLDPBD5394-5G BLDPBD5394-100G BLDPBD5394-1000G
trans-1,4-Cyclohexanedicarboxylic acid 97%
trans-1,4-Cyclohexanedicarboxylic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₁₂O₄
Molekylvægt: 172,18 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00066203
CAS-nummer: 619-82-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR