1,3,5-Trifluoro-2,4,6-triiodobenzene 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD53642-10GEA 807 DKK
BLDPBD53642-10G BLDPBD53642-250MG BLDPBD53642-100MG BLDPBD53642-100G BLDPBD53642-25G BLDPBD53642-1G BLDPBD53642-5G
1,3,5-Trifluoro-2,4,6-triiodobenzene 98%
1,3,5-Trifluoro-2,4,6-triiodobenzene

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₆F₃I₃
Molekylvægt: 509,77 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06248899
CAS-nummer: 84322-56-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR