2,4-Xylidine hydrochloride 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD5291-5GEA 303 DKK
BLDPBD5291-5G BLDPBD5291-1G BLDPBD5291-100G BLDPBD5291-25G
2,4-Xylidine hydrochloride 98%
2,4-Xylidine hydrochloride

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₁₂ClN
Molekylvægt: 157,6406 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00060209
CAS-nummer: 21436-96-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR