4-(Methylsulphonyl)phenylboronic acid 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD5036-10GEA 435 DKK
BLDPBD5036-10G BLDPBD5036-100G BLDPBD5036-25G BLDPBD5036-1G BLDPBD5036-5G
4-(Methylsulphonyl)phenylboronic acid 98%
4-(Methylsulphonyl)phenylboronic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₇H₉BO₄S
Molekylvægt: 200,02 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01630820
CAS-nummer: 149104-88-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR