4-(Ethylsulphonyl)benzaldehyde 97%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD496055-5GEA
4240
DKK
BLDPBD496055-5G
BLDPBD496055-1G
BLDPBD496055-250MG
BLDPBD496055-10G
4-(Ethylsulphonyl)benzaldehyde 97%
4-(Ethylsulphonyl)benzaldehyde
Storage condition: Inert atmosphere, 2-8°C
Formel:
C₉H₁₀O₃S Molekylvægt: 198,239 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD09264545 CAS-nummer: 50899-03-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...