3-Bromo-6-chloroisoquinoline 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD493294-100MGEA 1070 DKK
BLDPBD493294-100MG
3-Bromo-6-chloroisoquinoline 95%
3-Bromo-6-chloroisoquinoline

Storage condition: Inert atmosphere, 2-8°C

Formel: C₉H₅BrClN
Molekylvægt: 242,5 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD18793407
CAS-nummer: 1276056-79-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR