4-(4-Chlorophenyl)-3-thiosemicarbazide 98+%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD4928-5GEA 566 DKK
BLDPBD4928-5G BLDPBD4928-250MG BLDPBD4928-1G
4-(4-Chlorophenyl)-3-thiosemicarbazide 98+%
4-(4-Chlorophenyl)-3-thiosemicarbazide

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₇H₈ClN₃S
Molekylvægt: 201,68 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00041295
CAS-nummer: 22814-92-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR