(S)-3-Isopropylmorpholine 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD48952-1GEA 1420 DKK
BLDPBD48952-1G BLDPBD48952-250MG BLDPBD48952-100MG BLDPBD48952-5G
(S)-3-Isopropylmorpholine 98%
(S)-3-Isopropylmorpholine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₇H₁₅NO
Molekylvægt: 129,2 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11519112
CAS-nummer: 77897-21-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR