3,4-Diaminobenzonitrile 97%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD4805-5GEA
210
DKK
BLDPBD4805-5G
BLDPBD4805-25G
BLDPBD4805-10G
BLDPBD4805-100G
BLDPBD4805-1G
3,4-Diaminobenzonitrile 97%
3,4-Diaminobenzonitrile
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formel:
C₇H₇N₃ Molekylvægt: 133,15 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00723901 CAS-nummer: 17626-40-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...