(S)-(-)-1-Phenylethanol 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD48024-25GEA 334 DKK
BLDPBD48024-25G BLDPBD48024-5G BLDPBD48024-10G BLDPBD48024-1G
(S)-(-)-1-Phenylethanol 98%
(S)-(-)-1-Phenylethanol

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₁₀O
Molekylvægt: 122,17 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00064264
CAS-nummer: 1445-91-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR