4-Carboxyphenylboronic acid 98%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD4728-100GEA 559 DKK
BLDPBD4728-100G BLDPBD4728-25G BLDPBD4728-10G BLDPBD4728-1G BLDPBD4728-5G BLDPBD4728-500G
4-Carboxyphenylboronic acid 98%
4-Carboxyphenylboronic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₇H₇BO₄
Molekylvægt: 165,94 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00151801
CAS-nummer: 14047-29-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR