(R)-1-(Pyrrolidin-2-ylmethyl)pyrrolidine 95% (by GC)

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD46823-250MGEA 419 DKK
BLDPBD46823-250MG BLDPBD46823-1G
(R)-1-(Pyrrolidin-2-ylmethyl)pyrrolidine 95% (by GC)
(R)-1-(Pyrrolidin-2-ylmethyl)pyrrolidine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₉H₁₈N₂
Molekylvægt: 154,2500 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD04117879
CAS-nummer: 60419-23-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR